Order Entry
Austria
ContactUsLinkComponent
Dropping pipettes
Dropping pipettes
Catalog # HERE1090301
Supplier:  HERENZ HEINZ
undefined
Dropping pipettes
Catalog # HERE1090301
Supplier:  HERENZ HEINZ

Specifications

  • Description:
    Dropping pipettes, AR-glass, without bulb
  • Length:
    80 mm
  • Seq:
    01
  • Pk:
    1
  • Stem Ø:
    0,7 mm

Specifications

About this item

Dropping pipettes made from AR-Glas®.

  • Without bulb