Order Entry
Austria
ContactUsLinkComponent
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in 2-propanol/methanol, Titripur® Reag. Ph. Eur., Reag. USP for titration in non-aqueous media, Supelco®
Flammable
Corrosive
Health
Irritant
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in 2-propanol/methanol, Titripur® Reag. Ph. Eur., Reag. USP for titration in non-aqueous media, Supelco®
Supplier:  Merck
Certificates
Flammable
Corrosive
Health
Irritant
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in 2-propanol/methanol, Titripur® Reag. Ph. Eur., Reag. USP for titration in non-aqueous media, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Form:
    liquid
  • Amount-of-substance concentration:
    0.0994 - 0.1006
  • Measurement uncertainty:
    +/- 0.0004
  • Traceability:
    NIST SRM
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

Frequently Bought Together