Order Entry
Austria
ContactUsLinkComponent
 
Organic reference standard, 1-Methylfluoranthene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL20892500CY
 :  
Organic reference standard, 1-Methylfluoranthene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL20892500CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    25889-60-5
  • Description:
    1-Methylfluoranthene 10 µg/ml in cyclohexane
  • Formula:
    C₁₇H₁₂
  • Description:
    1-Methylfluoranthene