Organic reference standard, Benzo[k]fluoranthene 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA20580000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:207-08-9
- Description:Benzo[k]fluoranthene 100 µg/ml in cyclohexane
- Environmentally Preferable:
- Formula:C₂₀H₁₂
- Description:Benzo[k]fluoranthene