Organic reference standard, Benzo[k]fluoranthene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERL20580000AL
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:207-08-9
- Description:Benzo[k]fluoranthene 10 µg/ml in acetonitrile
- Formula:C₂₀H₁₂
- Description:Benzo[k]fluoranthene