Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Benzo[j]fluoranthene 2000 µg/ml in dichloromethane, Supelco®
Organic reference standard, Benzo[j]fluoranthene 2000 µg/ml in dichloromethane, Supelco®
Catalog # 502332.
Supplier:  Merck
CAS Number:  
Organic reference standard, Benzo[j]fluoranthene 2000 µg/ml in dichloromethane, Supelco®
Catalog # 502332.
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    205-82-3
  • Description:
    Benzo[j]fluoranthene 2,000 µg/ml in dichloromethane
  • Environmentally Preferable:
  • Formula:
    C₂₀H₁₂
  • Description:
    Benzo[j]fluoranthene

Specifications

About this item