Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Benzo[b]fluoranthene 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Organic reference standard, Benzo[b]fluoranthene 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA20565000AL
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Benzo[b]fluoranthene 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA20565000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    205-99-2
  • Description:
    Benzo[b]fluoranthene 100 µg/ml in acetonitrile
  • Environmentally Preferable:
  • Formula:
    C₂₀H₁₂
  • Description:
    Benzo[b]fluoranthene

Specifications

About this item