
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Catalog # ROCH10760994001
Supplier: ROCHE
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:433.6214 g/mol
:M-Clarity™ quality level = MQ100
EINECS:229-506-1
:GHS07
Storage Temperature:Freezer
CAS Number:6578-06-9
Formula:C₁₅H₁₅BrClN₂O₄P
MDL Number:MFCD00040636