
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Supplier: Biotium
Certificates
About this item
CAS Number:6578-06-9
:
Formula:C₁₅H₁₅BrClN₂O₄P
MDL Number:MFCD00040636
:433.6214 g/mol
EINECS:229-506-1
:GHS07
Storage Temperature:Freezer
Frequently Bought Together