Specifications
- Pack type:Plastic bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
:
:370.43 g/mol
CAS Number:102185-33-1
Formula:C₈H₄BrClNNa₂O₄P
MDL Number:MFCD00036757