Order Entry
Austria
ContactUsLinkComponent
 
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Catalog # MOLEM46642783
Supplier:  Molekula
CAS Number:  
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Catalog # MOLEM46642783
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
 :
 :370.43 g/mol
CAS Number:102185-33-1
Formula:C₈H₄BrClNNa₂O₄P
MDL Number:MFCD00036757