5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥99%, off-white powder
: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
:102185-33-1
:
:C₈H₄BrClNNa₂O₄P
:MFCD00036757
:370.43 g/mol
:>300 °C (lit.)
:Freezer
- Environmentally Preferable:
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Reference
- Loss on Drying:≤10%
- UV/VIS: I max (water):291 ± 3nm
Frequently Bought Together