Order Entry
Austria
ContactUsLinkComponent
 
8-Hydroxyquinoline-5-sulphonic acid monohydrate for synthesis, Sigma-Aldrich®
 :  Merck
 
8-Hydroxyquinoline-5-sulphonic acid monohydrate for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Environmentally Preferable:

 

 

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    322 °C
  • Molar Mass:
    243.24 g/mol
  • Hill Formula:
    C9H7NO4S*H2O

 

 

Frequently Bought Together