

8-Hydroxyquinoline-5-sulphonic acid monohydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:283158-18-9
:
Formula:C₉H₇NO₄S·1H₂O
MDL Number:MFCD00149482
:243.2365 g/mol
Melting Pt:>300 °C
EINECS:201-570-5
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Assay:≥97.5 %
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together