Order Entry
Austria
ContactUsLinkComponent
Digoxin
Digoxin
Catalog # EHERC12633850
Supplier:  EHRENSTORFER
CAS Number:  
Digoxin
Catalog # EHERC12633850
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :780.95 g/mol
EINECS:244-068-1
UN:2811
ADR:6.1,II
CAS Number:20830-75-5
Formula:C₄₁H₆₄O₁₄
MDL Number:MFCD00003674