Organic reference standard, Dibenz[a,h]anthracene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20700000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:53-70-3
- Description:Dibenz[a,h]anthracene 10 µg/ml in acetonitrile
- Formula:C₂₂H₁₄
- Description:Dibenz[a,h]anthracene