Order Entry
Austria
ContactUsLinkComponent
(4-Chlorophenoxy)acetonitrile ≥98%
Toxic
Irritant
(4-Chlorophenoxy)acetonitrile ≥98%
Catalog # A15389.14
CAS Number:  
Toxic
Irritant
(4-Chlorophenoxy)acetonitrile ≥98%
Catalog # A15389.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

CAS Number:3598-13-8
 :
Formula:C₈H₆ClNO
 :167.60 g/mol
Boiling Pt:149…150 °C (15 mmHg)
Melting Pt:45…49 °C
UN:3439
ADR:6.1,III
 :GHS06
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00031535