Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2,4-Dichlorophenoxy)acetonitrile
(2,4-Dichlorophenoxy)acetonitrile
(2,4-Dichlorophenoxy)acetonitrile
(2,4-Dichlorophenoxy)acetonitrile
MAYBSPB00370.50
:
Thermo Scientific
:
(2,4-Dichlorophenoxy)acetonitrile
MAYBSPB00370.50
:
Thermo Scientific
:
Pack type:
Glass bottle for solids
Pk:
50 g
Seq:
3368
:
:
202.04 g/mol
:
3439
:
6.1,III
:
3956-63-6
:
C₈H₅Cl₂NO
:
MFCD00019826