Order Entry
Austria
ContactUsLinkComponent
Pentachlorophenol
Toxic
Health
Environment
Pentachlorophenol
Catalog # ACCUA031
Supplier:  AccuStandard
CAS Number:  
Toxic
Health
Environment
Pentachlorophenol
Catalog # ACCUA031
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :266.34 g/mol
Merck Index:12,07242
Boiling Pt:310 °C (1013 hPa)
Melting Pt:188 °C
Density:1.98 g/cm³ (20 °C)
EINECS:201-778-6
UN:3155
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
CAS Number:87-86-5
Formula:C₆HCl₅O