Order Entry
Austria
ContactUsLinkComponent
2-Propanol VLSI for microelectronic, J.T.Baker®
2-Propanol VLSI for microelectronic, J.T.Baker®
Catalog # 5373-05
Supplier:  Avantor
CAS Number:  
undefined
2-Propanol VLSI for microelectronic, J.T.Baker®
Catalog # 5373-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    8 pt US
  • Seq:
    8026

Specifications

Specification Test Results

  • For Microelectronic Use:
    = 99.5 %
  • Assay (CH₃CHOHCH₃):
    ≥ 99.5 %
  • Color (APHA):
    ≤ 10
  • Residue after Evaporation:
    ≤ 4 ppm
  • Solubility in H₂O:
    Passes Test
  • Water (H₂O)(by Karl Fischer titrn):
    ≤ 0.05 %
  • Acidity (µeq/g):
    ≤ 0.2
  • Alkalinity (µeq/g):
    ≤ 0.1
  • Chloride (Cl):
    ≤ 0.2 ppm
  • Phosphate (PO₄):
    ≤ 0.3 ppm
  • Trace Impurities - Aluminum (Al):
    ≤ 50.0 ppb
  • Arsenic and Antimony (as As):
    ≤ 10.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤ 20.0 ppb
  • Trace Impurities - Boron (B):
    ≤ 10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤ 20.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤ 50.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤ 20.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤ 20 ppb
  • Trace Impurities - Copper (Cu):
    ≤ 10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤ 50 ppb
  • Trace Impurities - Germanium (Ge):
    ≤ 50.0 ppb
  • Trace Impurities - Gold (Au):
    ≤ 20 ppb
  • Heavy Metals (as Pb):
    ≤ 200.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤ 50.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤ 20.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤ 50.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤ 20 ppb
  • Trace Impurities - Manganese (Mn):
    ≤ 20.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤ 10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤ 100 ppb
  • Trace Impurities - Silicon (Si):
    ≤ 50 ppb
  • Trace Impurities - Silver (Ag):
    ≤ 20.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤ 100.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤ 20.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤ 100 ppb
  • Trace Impurities - Titanium (Ti):
    ≤ 20 ppb
  • Trace Impurities - Zinc (Zn):
    ≤ 50 ppb
  • Particle Count - 0.5 µm and greater:
    ≤ 100 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤ 8 par/ml

Specification Test Results

About this item

 :
 :60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
 :Management of Change (MOC) category = R
REACH:01-2119457558-25
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674