Order Entry
Austria
ContactUsLinkComponent
 
Flammable
Irritant
Acetone VLSI for microelectronic, J.T.Baker®
Catalog # 5356-05
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
Acetone VLSI for microelectronic, J.T.Baker®
Catalog # 5356-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    8 pt US
  • Seq:
    8026

Specifications

Specification Test Results

  • For Microelectronic Use:
    = 99.5 %
  • Assay ((CH₃)₂CO) (by GC, corrected for water):
    ≥99.5 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤5 ppm
  • Acidity (µeq/g):
    ≤0.3
  • Alkalinity (µeq/g):
    ≤0.5
  • Water (H₂O):
    ≤0.5 %
  • Solubility in H₂O:
    Passes Test
  • Chloride (Cl):
    ≤0.2 ppm
  • Phosphate (PO₄):
    ≤0.05 ppm
  • Trace Impurities - Aluminum (Al):
    ≤50.0 ppb
  • Arsenic and Antimony (as As):
    ≤5.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤20.0 ppb
  • Trace Impurities - Boron (B):
    ≤20.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤25.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤20.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤20 ppb
  • Trace Impurities - Iron (Fe):
    ≤20.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤20.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤50.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤20 ppb
  • Trace Impurities - Manganese (Mn):
    ≤10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤50 ppb
  • Trace Impurities - Sodium (Na):
    ≤50.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤50 ppb
  • Trace Impurities - Titanium (Ti):
    ≤20 ppb
  • Trace Impurities - Zinc (Zn):
    ≤50 ppb
  • Particle Count - 0.5 µm and greater:
    ≤60 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤8 par/ml

Specification Test Results

About this item

 :
 :58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
 :GHS02
 :GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765