Utility Cart
Supplier: Rubbermaid Commercial Products
Certificates
About this item
Trolleys are made of PP with four swivel castors.
- Easy to clean, smooth surfaces
- Plastic construction help to reduce noise
- Ergonomic, easy grip handles
- Quiet and large 102 mm non marking swivel castors
Load capacity: 90,7 kg
Related Information
Product Information
Frequently Bought Together