Order Entry
Austria
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP) ≥99.5% for peptide synthesis
N-Methyl-2-pyrrolidone (NMP) ≥99.5% for peptide synthesis
Catalog # 84572.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
N-Methyl-2-pyrrolidone (NMP) ≥99.5% for peptide synthesis
Catalog # 84572.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.5 %
  • Appearance:
    Clear colourless liquid
  • Colour value:
    Max. 10 APHA
  • Free amines:
    Max. 0.001 %
  • Residue on evaporation:
    Max. 0.002 %
  • Water:
    Max. 0.02 %
  • Fe (Iron):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Pb (Lead):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.1 ppm

Specification Test Results

About this item

CAS Number:872-50-4
 :
Formula:C₅H₉NO
MDL Number:MFCD00003193
 :99.13 g/mol
Merck Index:13,06140
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
Flash Pt:91 °C
Storage Temperature:Ambient
EINECS:212-828-1
REACH:01-2119472430-46
 :GHS08
 :GHS07