


N,N-Dimethylformamide ≥99.8% for peptide synthesis
Catalog # 84571.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (calculated on anhydrous):Min. 99.8 %
- Appearance:Clear colourless liquid
- Acidity:Max. 0.001 %
- Colour value:Max. 10 APHA
- Free amines:Max. 0.001 %
- Formaldehyde:Max. 0.002 %
- Residue on evaporation:Max. 0.001 %
- Water:Max. 0.01 %
- Fe (Iron):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Pb (Lead):Max. 0.1 ppm
- Zn (Zinc):Max. 0.1 ppm
Specification Test Results
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:−60.5 °C
Melting Pt:–60.5 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (DIN 51755) (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07