Order Entry
Austria
ContactUsLinkComponent
 
Plant Genomic DNA purification Mini kit, GeneJET™
 
Plant Genomic DNA purification Mini kit, GeneJET™

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Description:
    GeneJET™ Plant Genomic DNA Purification Mini Kit

 

Frequently Bought Together