N,N-Dimethylformamide for peptide synthesis, Supelco®
: Merck
: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
:68-12-2
:
:HCON(CH₃)₂
:MFCD00003284
:73.09 g/mol
:13,03269
:153 °C (1013 hPa)
:–61 °C
:0.949 g/cm³ (20 °C)
:Ambient
:200-679-5
:2265
:3,III
:M-Clarity™ quality level = MQ200
:57.5 °C (closed cup)
:01-2119475605-32
:GHS02
:GHS08
:GHS07
- Environmentally Preferable:
- Purity (GC):≥ 99.9 %
- Identity (IR):conforms
- Color:≤ 10 Hazen
- Acidity:≤ 0.0005 meq/g
- Alkalinity:≤ 0.0002 meq/g
- Amines (Dimethylamine):≤ 10 ppm
- Evaporation residue:≤ 10
- Water:≤ 300 ppm
- Density:0.94 g/cm3 (20 °C)
- pH:7 (200 g/l, H2O, 20 °C)
- Ignition Point:410 °C
Frequently Bought Together