Order Entry
Austria
ContactUsLinkComponent
Whole blood RNA purification mini kit, GeneJET™
Whole blood RNA purification mini kit, GeneJET™
Catalog # K0761
CAS Number:  
undefined
Whole blood RNA purification mini kit, GeneJET™
Catalog # K0761
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Blood
  • Target molecule:
    RNA
  • Description:
    GeneJET™ Whole Blood RNA Purification Mini Kit, 50 reactions
  • Description:
    GeneJET™ Whole Blood RNA Purification Mini Kit

Specifications

About this item

GeneJET™ Whole Blood RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from whole blood and related body fluids. The kit utilises silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 15 minutes. The purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting, and other RNA-based analysis.

  • Efficient: Up to 1,8 µg of total RNA from 500 µl of human blood
  • Convenient: Spin columns are capped and assembled with collection tubes
  • Pure: RNA pure from enzyme inhibitors, proteins, ready to use in all demanding applications; validated in cDNA synthesis and quantitative RT-PCR
  • Fast and simple: Procedure takes less than 15 minutes and does not involve organic extraction or differential RBC lysis
  • Compatible: Suitable for wide variety of samples, including human and animal blood treated with EDTA, citrate or heparin, buffy coat, bone marrow, and related body fluids
Delivery: Each kit includes lysis buffer, Wash Buffer 1 (concentrated), Wash Buffer 2 (concentrated), water (nuclease-free), GeneJET™ RNA purification columns (pre-assembled with collection tubes), and collection tubes (1,5 and 2 ml).