Order Entry
Austria
Orders LinkContactUsLinkComponent
Plasmid Maxiprep kit, GeneJET™

Plasmid Maxiprep kit, GeneJET™

Plasmid Maxiprep kit, GeneJET™
Certificates
Plasmid Maxiprep kit, GeneJET™

2 Options Available Below

Product Details & Documents

GeneJET™ Plasmid Maxiprep Kit is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures. GeneJET™ Plasmid Maxiprep Kit utilises silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 750 µg of high copy plasmid DNA that can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

  • Efficient: High yields up to 750 µg of high quality plasmid DNA from 250 ml of bacterial culture
  • High concentration of DNA purified: >400 ng/µl
  • Convenient: Spin column format, centrifuge or vacuum protocols
  • Fast: Procedure takes 60 minutes, considerably faster than using gravity flow columns

Delivery information: Each kit includes resuspension solution, lysis solution, neutralisation solution, endotoxin binding reagent, Wash Solution I (concentrated), Wash Solution II (concentrated), RNase A solution, elution buffer (10 mM Tris-HCl, pH 8,5), GeneJET™ Maxi purification columns pre-assembled with collection tubes (50 ml), and collection tubes (50 ml).

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • K0491
  • K0492
  • Description:
  • GeneJET™ Plasmid Maxiprep Kit, 10 preps
  • GeneJET™ Plasmid Maxiprep Kit, 25 preps
  • Target molecule:
  • Plasmid
  • Plasmid
  • Refine target molecule:
  • Plasmid
  • Plasmid
  • Sample type:
  • Bacteria
  • Bacteria
  • Method / Format:
  • Column (silica-based)
  • Column (silica-based)
  • Prep size:
  • Maxi
  • Maxi

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • K0491
    GeneJET™ Plasmid Maxiprep Kit, 10 preps
    GeneJET™ Plasmid Maxiprep Kit, 10 preps
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 190,00 €
    190,00 €
    1 * 1 KIT
       190,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    K0491
    Description
    GeneJET™ Plasmid Maxiprep Kit, 10 preps
    Target molecule
    Plasmid
    Refine target molecule
    Plasmid
    Sample type
    Bacteria
    Method / Format
    Column (silica-based)
    Prep size
    Maxi
  • K0492
    GeneJET™ Plasmid Maxiprep Kit, 25 preps
    GeneJET™ Plasmid Maxiprep Kit, 25 preps
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 440,00 €
    440,00 €
    1 * 1 KIT
       440,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    K0492
    Description
    GeneJET™ Plasmid Maxiprep Kit, 25 preps
    Target molecule
    Plasmid
    Refine target molecule
    Plasmid
    Sample type
    Bacteria
    Method / Format
    Column (silica-based)
    Prep size
    Maxi