ContactUsLinkComponent
Swabs with transport medium
  LPIT118898
undefined
Swabs with transport medium
  LPIT118898
  • Description:
    Swabs with transport medium
  • Shaft/head:
    Plastic/rayon
  • Transport medium:
    Stuart gel
  • Pk:
    250
Swabs with transport medium.
Specially designed for common applications.
 : Individually wrapped in a peel-pouch.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904