Order Entry
Austria
ContactUsLinkComponent
Shelving systems, solid shelves, MetroMax i®
Shelving systems, solid shelves, MetroMax i®
Catalog # METOX336EFX3
undefined
Shelving systems, solid shelves, MetroMax i®
Catalog # METOX336EFX3

Specifications

  • Description:
    Shelf system, solid shelves, 914×457 mm
  • Pk:
    1
  • Weight:
    42 kg

Specifications

About this item

Corrosion proof shelves and posts with removable polymer mats.

  • Consist of four open grid shelves and posts, wedge connectors and polyurethane casters (2-swivel and 2-brake)
  • Models available with antimicrobial product protection built into the tread
  • Maximum load capacity is upto 408 kg