Order Entry
Austria
ContactUsLinkComponent
Shelving systems, open grid, MetroMax i®
Shelving systems, open grid, MetroMax i®
Catalog # METOX566EGX3
undefined
Shelving systems, open grid, MetroMax i®
Catalog # METOX566EGX3

Specifications

  • Description:
    Shelf system, open grid,1524×610 mm
  • Pk:
    1
  • Weight:
    51 kg

Specifications

About this item

Corrosion proof shelves and posts with removable polymer mats.

  • Consist of four open grid shelves and posts, wedge connectors and polyurethane casters (2-swivel and 2-brake)
  • Microban® antimicrobial product protection is built into every shelf mat and touch points
  • Shelving up to 900 lbs. (408 kg) per shelf