Divider tote box for shelving systems, Metro®
Supplier: Metro International
New Product
Certificates
About this item
Divider tote boxes made of polypropylene base resins offer safe and efficient storage of items in bulk, or in compartmentalised fashion with the use of optional dividers and covers. Available in natural gray (NAT), Benstat™ blue static dissipate (BAS) or Bentron™ black conductive (CAS).
Frequently Bought Together