Mixing measuring cylinders, tall form, class A, borosilicate glass, FORTUNA®
Supplier: Poulten & Graf
Certificates
About this item
Borosilicate glass 3,3 class A, conformity certified.
- Calibrated to contain (TC, In)
- Hexagonal glass base, PP stopper
- Blue graduations
Certifications: DIN EN ISO 4788
Frequently Bought Together