Spray bottles, turn 'n' spray, LaboPlast®
Lieferant: BURKLE
Zertifikate
Über diesen Artikel
LDPE, translucent, PP spray attachment, PE tubing, stainless steel weight.
- Uninterrupted spraying, even when inverted
- Adjustable nozzle, diameter 0,6 mm
- Amount sprayed per stroke: 1,2 ml ±0,1
Spraying upside down is not possible with standard spray bottles. As soon as the suction tube is no longer immersed in liquid, it cannot spray. Not so with the overhead spray bottle Turn 'n' Spray. It features a special technology that allows you to spray from any position. That means it can spray practically and conveniently from any direction and reach hard to access places easily and without straining.
Frequently Bought Together