Order Entry
Austria
Orders LinkContactUsLinkComponent
RevertAid™ H Minus Reverse Transcriptase | Avantor

RevertAid™ H Minus Reverse Transcriptase

RevertAid™ H Minus Reverse Transcriptase
Certificates
RevertAid™ H Minus Reverse Transcriptase
RevertAid™ H Minus Reverse Transcriptase

2 Options Available Below

Product Details & Documents

RevertAid™ H Minus Reverse Transcriptase (RT) is a genetically modified M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity, but lacks RNase H activity due to point mutation in the RNase H domain. RevertAid™ H Minus Reverse Transcriptase does not degrade RNA in RNA-DNA hybrids during synthesis of the first strand cDNA and therefore high yields of full-length cDNA from long templates are obtained. RevertAid™ H Minus Reverse Transcriptase maintains activity over a wide temperature range (42 to 55 °C), which makes the enzyme an ideal tool for reverse transcription of RNAs having a high degree of secondary structure.

  • High yields of full length first strand cDNA up to 13 kb
  • Optimum activity at 42 to 45 °C
  • Active up to 55 °C
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, fluorescein-labelled nucleotides)

Delivery information: Supplied with 5X reaction buffer.

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • EP0451
    RevertAid™ H Minus Reverse Transcriptase, 10 000 units (200 U/µl), for 50 reactions of 20 µl
    RevertAid™ H Minus Reverse Transcriptase, 10 000 units (200 U/µl), for 50 reactions of 20 µl
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 214,00 €
    214,00 €
    1 * 1 KIT
       214,00 €

  • EP0452
    RevertAid™ H Minus Reverse Transcriptase, 5×10 000 units (200 U/µl), for 250 reactions of 20 µl
    RevertAid™ H Minus Reverse Transcriptase, 5×10 000 units (200 U/µl), for 250 reactions of 20 µl
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 812,00 €
    812,00 €
    1 * 1 KIT
       812,00 €

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.