DreamTaq™ DNA polymerase
DreamTaq™ DNA Polymerase is an enhanced Taq DNA polymerase optimised for all standard PCR applications. It ensures higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase. DreamTaq™ DNA Polymerase uses the same reaction set-up and cycling conditions as conventional Taq DNA polymerase. Extensive optimisation of reaction conditions is not required. The enzyme is inhibited by dUTP but can incorporate modified nucleotides. Applications include routine PCR amplification of DNA fragments up to 6 kb from genomic DNA and up to 20 kb from viral DNA, RT-PCR, and generation of PCR products for TA cloning.
- Robust amplification with minimal optimisation
- High yields of PCR products
- Higher sensitivity compared to conventional Taq DNA polymerase
- Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
- Generates 3’-dA overhangs
- Incorporates modified nucleotides
: Supplied with 10X DreamTaq™ buffer, which includes 20 mM MgCl₂.
Frequently Bought Together