ContactUsLinkComponent
Analogue stopwatch, Lumen
Analogue stopwatch, Lumen
  609-1115
 :  HANHART
undefined
Analogue stopwatch, Lumen
  609-1115
 :  HANHART
  • Ø:
    55 mm
  • Description:
    1/5 s, 30 min, 80 g
  • Pk:
    1
These watches, with luminous dials, are ideal for use in industry, laboratories and for athletes. Once the special dial has been 'charged' with light (takes approx. 1 hour), the face will glow in the dark for approx. 3 to 4 hours so it is clearly readable. This requires no additional energy or radiating materials. This is achieved by using an innovative plastic material containing light-storing pigments.
  • Robust ABS case with additional dust and water protection
  • Glass and glass fixing ring are moulded together and then firmly screwed to the case
  • Pin lever, 1 jewel