ContactUsLinkComponent
Cleanroom face masks, BioClean Microflow™
Cleanroom face masks, BioClean Microflow™
Catalog # 113-8158
Supplier:  Nitritex
undefined
Cleanroom face masks, BioClean Microflow™
Catalog # 113-8158
Supplier:  Nitritex
Specifications
  • Type:
    Non sterile, with head loop
  • Colour:
    White
  • Size:
    375×330 mm
  • Pk:
    1.000
Specifications
About this item
Non sterile soft face veil with headband and nosepiece manufactured using ultrasonic bonding.
  • Veil made from low linting thermabond non woven material
  • White hydroentangled polyester binding and tubular polyester head loop
  • Fully enclosed, flexible, malleable aluminium nosepiece coated with plastic
  • Processed in a cleanroom environment, contains no latex
Suitable for use in class 10, ISO Class 4, M2.5 cleanrooms.
Certifications: Manufactured in a facility holding ISO 9001:2008. Processed in an NEBB certified ISO class 4 cleanroom environment. Sampling and inspection in accordance with ISO 2859-3:1994.
Packaging: 100 veils per double bag; 10 bags per case (1000 veils).