Orders LinkContactUsLinkComponent
Sample Spoons, Disposable, SteriWare®
Sample Spoons, Disposable, SteriWare®
Sample Spoons, Disposable, SteriWare®
Supplier:  Sampling Systems
Certificates
Sample Spoons, Disposable, SteriWare®
Sample Spoons, Disposable, SteriWare®
7 Options Available Below
Product Details & Documents
These disposable, sterile sample spoons are ideal for dispensing and sampling small quantities. Manufactured from food grade polystyrene (PS) or polyproylene (PP).
  • Moulded and packed in a cleanroom
  • Available in white, red and blue
  • Made from FDA and EU conforming polymers
  • BSE/TSE-free
  • Available as gamma irradiated sterile (SAL 10⁻⁶)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
Full batch traceability.
Packaging: Individually bagged.
Product Family Options
Product Information
  • Length
    Spoon size
    Capacity
    Material
    Availability
    Price
  • White, non sterile
    173 mm
    26×25 mm
    3 ml
    PS
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 99,70 €
    99,70 €
    1 * 100 ST
       99,70 €
  • White, sterile
    173 mm
    26×25 mm
    3 ml
    PS
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 140,00 €
    140,00 €
    1 * 100 ST
       140,00 €
  • White, non sterile
    185 mm
    48×35 mm
    10 ml
    PS
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 99,70 €
    99,70 €
    1 * 100 ST
       99,70 €
  • White, sterile
    185 mm
    48×35 mm
    10 ml
    PS
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 140,00 €
    140,00 €
    1 * 100 ST
       140,00 €
  • SAMP8088B-10
    Blue, non sterile
    Blue, non sterile
    185 mm
    48×35 mm
    10 ml
    PP
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 110,00 €
    110,00 €
    1 * 100 ST
       110,00 €
  • Blue, sterile
    185 mm
    48×35 mm
    10 ml
    PP
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 142,00 €
    142,00 €
    1 * 100 ST
       142,00 €
  • SAMP8088R-10S
    Red, sterile
    Red, sterile
    185 mm
    48×35 mm
    10 ml
    PS
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 147,00 €
    147,00 €
    1 * 100 ST
       147,00 €