Anchorage lines, MH00
Twisted polyamide rope suitable for 14 mm/16 mm rope grabs. Suitable for vertical, horizontal and sloping applications.
- Reliable and lightweight
- Breaking strength: >22 KN
: EN 353-2: Guide type fall arresters on a flexible anchorage line Comply with the standard if used with MILLER rope grab only; EN 362 (rope)
: Supplied with a counterweight and screwgate karabiner ML00 complies with EN696
- Catalog No:
- 118-0425
- 118-0422
- 118-0423
- 118-0421
- Material:
- Polyamide
- Polyamide
- Polyamide
- Polyamide
Frequently Bought Together