Flasks acc. to Kjeldahl, with beaded rim
WITG5.518.001
- Ext.Ø:131 mm
- Capacity:1000 ml
- Height:350 mm
- Neck Ø ext.:34 mm
- Pk:1
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With printed indication of contents and white labelling area.