Orders LinkContactUsLinkComponent
Trypsinising flasks
Trypsinising flasks
Supplier:   witeg Labortechnik
Trypsinising flasks
Supplier:  witeg Labortechnik
Certificates
Trypsinising flasks
Trypsinising flasks
6 Options Available Below
Product Details & Documents
Trypsinising flasks, graduated, designed for converting tissue fragments and homogenates into cell suspensions by trypsin digestion of connective proteins. Flutes in flask enhance vigorous agitation.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,10 €
    44,10 €
    1 * 1 ST
       44,10 €
  • 500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 48,60 €
    48,60 €
    1 * 1 ST
       48,60 €
  • 1000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,80 €
    59,80 €
    1 * 1 ST
       59,80 €
  • 2000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,90 €
    86,90 €
    1 * 1 ST
       86,90 €
  • 3000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 149,00 €
    149,00 €
    1 * 1 ST
       149,00 €
  • 5000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 224,00 €
    224,00 €
    1 * 1 ST
       224,00 €