Order Entry
Austria
ContactUsLinkComponent
HF perfusion filters, CultureGard®
HF perfusion filters, CultureGard®
Catalog # SPECCU2M20512N
HF perfusion filters, CultureGard®
Catalog # SPECCU2M20512N

Specifications

  • Filtration area:
    70 cm²
  • Pore size:
    0,2 µm
  • Sterile:
    -
  • Capacity:
    2 – 50 l
  • Pk:
    12

Specifications

About this item

CultureGard® HF perfusion hollow fibre sterilising grade filter for cell culture perfusion. Made with 0,2 µm MWCO DynaFibre® membrane that permits the flow of culture medium while preventing the introduction of foreign microbials.

  • These specialised filters have a large surface area for faster flow rates under lower pressures
  • Lower pressure during filtration reduces the risk of the membrane clogging or rupturing
  • The DynaFibre® membrane is hydrophilic so there is minimum hold-up volume
  • PP vent fibre prevents air lock
  • Inlet/outlet connection: Female luer-Lok® (FLL) /male luer-Lok (MLL)

The combination of faster flow rates and minimum hold-up volume makes CultureGard® the ideal filters for adding even the most viscous nutrients, like 6 M glucose or media with serum, to hollow fibre or stirred tank bioreactors. Nutrients can be easily added to a culture without a syringe and septum.