Order Entry
Austria
ContactUsLinkComponent
Test tubes without rim, heavy wall
Test tubes without rim, heavy wall
Catalog # WITG4.904.002
Supplier:  witeg Labortechnik
undefined
Test tubes without rim, heavy wall
Catalog # WITG4.904.002
Supplier:  witeg Labortechnik

Specifications

  • Ext.Ø:
    8 mm
  • Wall thickness:
    1 mm
  • Length:
    40 mm
  • Pk:
    200

Specifications

About this item

AR-Glass, clear.

  • Excellent chemical resistance
  • Good temperature resistance