Culture tubes
Catalog # WITG4.903.101
Supplier: witeg Labortechnik
Specifications
- Ø:12 mm
- Wall thickness:0,8 - 1,0 mm
- Height:75 mm
- Pk:100
Specifications
About this item
Borosilicate glass 3,3 with rim.
- Excellent chemical resistance
- Excellent temperature resistance