Orders LinkContactUsLinkComponent
Cuvettes for macrophotometers | Avantor
Cuvettes for macrophotometers
Cuvettes for macrophotometers
Supplier:  LP ITALIANA
Certificates
Cuvettes for macrophotometers
Cuvettes for macrophotometers
2 Options Available Below
Product Details & Documents
Polystyrene cuvettes specially designed for macrophotometers.
Dokumente
Product Family Options
Product Information
  • Height
    Volume
    Path length
    Window material
    Availability
    Price
  • 45 mm
    2 ml
    10 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 163,00 €
    163,00 €
    1 * 1.000 ST
       163,00 €
  • 45 mm
    4 ml
    10 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 153,00 €
    153,00 €
    1 * 1.000 ST
       153,00 €