Orders LinkContactUsLinkComponent
Semi-solid samplers, MaxiCorer | Avantor
Semi-solid samplers, MaxiCorer
Supplier:   Sampling Systems
Semi-solid samplers, MaxiCorer
Supplier:  Sampling Systems
Certificates
Semi-solid samplers, MaxiCorer
Semi-solid samplers, MaxiCorer
2 Options Available Below
Product Details & Documents
The MaxiCorer can be used for sampling or dispensing small quantities of products. This designed for powders and granules, as well as for semi-solids such as cheese and thick grease in production, quality control or laboratories.
  • Fast and efficent core sampler
  • Long and curved blade construction
  • Hygienic design that can be easily cleaned
Product Family Options
Product Information
  • Material
    Length
    Immersion depth
    Ø
    Availability
    Price
  • SAMP7010A-01
    Semi-solid sampler
    Semi-solid sampler
    Stainless steel 316
    300 mm
    200 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 167,00 €
    167,00 €
    1 * 1 ST
       167,00 €
  • SAMP7010A-02
    Semi-solid sampler
    Semi-solid sampler
    Stainless steel 316
    600 mm
    500 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 204,00 €
    204,00 €
    1 * 1 ST
       204,00 €