Orders LinkContactUsLinkComponent
Viscous samplers
Viscous samplers
Supplier:   Sampling Systems
Viscous samplers
Supplier:  Sampling Systems
Certificates
Viscous samplers
Viscous samplers
3 Options Available Below
Product Details & Documents
These viscous samplers are designed for creams, shampoos, soaps, honey and other semi-viscous materials.
After sampling, the sampler can be completely dismantled, for a quick and thorough cleaning.
Product Family Options
Product Information
  • Material
    Capacity
    Length
    Availability
    Price
  • 231-0487
    Viscous sampler for medium viscosity materials
    Viscous sampler for medium viscosity materials
    Stainless steel 316
    300 ml
    1000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 010,00 €
    1 010,00 €
    1 * 1 ST
       1 010,00 €
  • SAMP7000S-1500
    Viscous sampler
    Viscous sampler
    Stainless steel 316L
    450 ml
    1500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 140,00 €
    1 140,00 €
    1 * 1 ST
       1 140,00 €
  • SAMP7000S-600
    Viscous sampler
    Viscous sampler
    Stainless steel 316L
    160 ml
    600 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 907,00 €
    907,00 €
    1 * 1 ST
       907,00 €