Orders LinkContactUsLinkComponent
Separating funnels, conical
Separating funnels, conical
Supplier:   witeg Labortechnik
Separating funnels, conical
Supplier:  witeg Labortechnik
Certificates
Separating funnels, conical
Separating funnels, conical
3 Options Available Below
Product Details & Documents
Separating funnels, conical, made of DURAN® borosilicate glass 3.3, graduated, with Witaflo-needle valve stopcock and PTFE-needle spindle.
  • Easy to read graduation
  • With plastic stopper
  • Very good chemical and temperature resistance
Product Family Options
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 1000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 235,00 €
    235,00 €
    1 * 2 ST
       235,00 €
  • 2000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 171,00 €
    171,00 €
    1 * 1 ST
       171,00 €
  • 500 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 176,00 €
    176,00 €
    1 * 2 ST
       176,00 €