Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
Supplier:   witeg Labortechnik
Separating funnels, Squibb
Supplier:  witeg Labortechnik
Certificates
Separating funnels, Squibb
Separating funnels, Squibb
2 Options Available Below
Product Details & Documents
Separatory funnels, Squibb, made of borosilicate glass 3.3, stopcock with economy PTFE-needle valve.
  • Easy to read graduation
  • Very good chemical resistance
  • Very good temperature resistance
Product Family Options
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 250 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 67,20 €
    67,20 €
    1 * 1 ST
       67,20 €
  • 500 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,50 €
    70,50 €
    1 * 1 ST
       70,50 €